Learn More
Grainger End Mill Kyocera MECXL 1000S100075T 1.
Catalog No. 502764757
Encompass_Preferred
The Kyocera 170HN8 is a 1/2" diameter, 4 flute, high speed steel end mill with a 1-1/2" overall length. It has a TiAlN coating for increased hardness and heat resistance. The end mill has a 30 degree helix angle and can be run at speeds up to 325 SFM. It is designed for slotting, profiling, contouring, and face milling in both ferrous and non-ferrous materials. The end mill features Kyocera's high precision grinding to ensure tight tolerances.
Catalog No. 50-276-4757 Supplier Grainger Supplier No. 170HN8
The Kyocera 170HN8 is a 1/2" diameter, 4 flute, high speed steel end mill with a 1-1/2" overall length. It has a TiAlN coating for increased hardness and heat resistance. The end mill has a 30 degree helix angle and can be run at speeds up to 325 SFM. It is designed for slotting, profiling, contouring, and face milling in both ferrous and non-ferrous materials. The end mill features Kyocera's high precision grinding to ensure tight tolerances.