Learn More
Grainger End Mill Kyocera MEW 0625W500102T 0.62
Catalog No. 502764759
Encompass_Preferred
The Kyocera 170HN8 is a 1/2 inch diameter, 4 flute, high speed steel end mill with a 1-1/2 inch overall length. It has a TiAlN coating for increased hardness and heat resistance. The end mill has a 30 degree helix angle and can be run at speeds up to 325 SFM. It is designed for slotting, profiling, and contouring in steels and cast irons. The end mill features Kyocera's KY2000 coating for high abrasion resistance.
Catalog No. 50-276-4759 Supplier Grainger Supplier No. 170HH8
The Kyocera 170HN8 is a 1/2 inch diameter, 4 flute, high speed steel end mill with a 1-1/2 inch overall length. It has a TiAlN coating for increased hardness and heat resistance. The end mill has a 30 degree helix angle and can be run at speeds up to 325 SFM. It is designed for slotting, profiling, and contouring in steels and cast irons. The end mill features Kyocera's KY2000 coating for high abrasion resistance.