Learn More
Grainger End Mill Kyocera MEW 0625W625102T 0.62
Catalog No. 502764360
Encompass_Preferred
The Kyocera 170HH9 is a 2 flute, high speed steel end mill with a 1/2" shank diameter, 1-1/2" overall length, and 3/4" cutting diameter. It has a TiAIN coating for wear resistance when machining steel, cast iron, alloy steels, stainless steels, titanium alloys, nickel alloys, and other non-ferrous metals. The end mill features a 30 degree helix angle and can achieve surface speeds up to 325 SFM. It is made in Japan.
Catalog No. 50-276-4360 Supplier Grainger Supplier No. 170HH9
The Kyocera 170HH9 is a 2 flute, high speed steel end mill with a 1/2" shank diameter, 1-1/2" overall length, and 3/4" cutting diameter. It has a TiAIN coating for wear resistance when machining steel, cast iron, alloy steels, stainless steels, titanium alloys, nickel alloys, and other non-ferrous metals. The end mill features a 30 degree helix angle and can achieve surface speeds up to 325 SFM. It is made in Japan.