Learn More
Grainger End Mill Kyocera MEW 0750W7504103T 0.7
Catalog No. 502764359
Encompass_Preferred
The Kyocera 170HK4 is a 4 flute, high performance carbide end mill with a 1/2" shank diameter. It has a 1" cutting diameter and can cut up to 1-1/4" deep. The overall length is 3-1/2". This end mill is designed for high feed milling of hardened steels up to 62 HRC, tool steels, mold steels, die steels, ADI, and nodular cast iron. Its advanced substrate and coating deliver high metal removal rates. The 170HK4 features a variable helix flute geometry to reduce harmonics and provide excellent chip control.
Catalog No. 50-276-4359 Supplier Grainger Supplier No. 170HK4
The Kyocera 170HK4 is a 4 flute, high performance carbide end mill with a 1/2" shank diameter. It has a 1" cutting diameter and can cut up to 1-1/4" deep. The overall length is 3-1/2". This end mill is designed for high feed milling of hardened steels up to 62 HRC, tool steels, mold steels, die steels, ADI, and nodular cast iron. Its advanced substrate and coating deliver high metal removal rates. The 170HK4 features a variable helix flute geometry to reduce harmonics and provide excellent chip control.