Learn More
Grainger End Mill Kyocera MEW 1000W10045103T 1.
Catalog No. 502763835
Encompass_Preferred
The Kyocera 170HK7 is a 1/2" diameter, 4 flute, high speed steel end mill with a 1-1/2" overall length. It has a TiAIN coating for increased hardness and lubricity. The end mill has a 30 degree helix angle and can be run at speeds up to 325 SFM. It is designed for slotting, profiling, contouring, and face milling in both ferrous and non-ferrous materials. The end mill features a center cutting design for plunging.
Catalog No. 50-276-3835 Supplier Grainger Supplier No. 170HK7
The Kyocera 170HK7 is a 1/2" diameter, 4 flute, high speed steel end mill with a 1-1/2" overall length. It has a TiAIN coating for increased hardness and lubricity. The end mill has a 30 degree helix angle and can be run at speeds up to 325 SFM. It is designed for slotting, profiling, contouring, and face milling in both ferrous and non-ferrous materials. The end mill features a center cutting design for plunging.