Learn More
Grainger End Mill Kyocera MEW 28S25103T 28mm
Catalog No. 502764770
Encompass_Preferred
The Kyocera 170HU0 is a 1/2" diameter, 4 flute, uncoated solid carbide end mill with a 1-1/2" overall length. It has a 30 degree helix angle and is center cutting. The end mill has a 0.500" shank diameter and is designed for high speed machining in steel, hardened steel, stainless steel, titanium, inconel, waspaloy, nimonic, and cast iron. It can be run at speeds up to 400 SFM. The end mill is made in Japan by Kyocera and is sold individually.
Catalog No. 50-276-4770 Supplier Grainger Supplier No. 170HU0
The Kyocera 170HU0 is a 1/2" diameter, 4 flute, uncoated solid carbide end mill with a 1-1/2" overall length. It has a 30 degree helix angle and is center cutting. The end mill has a 0.500" shank diameter and is designed for high speed machining in steel, hardened steel, stainless steel, titanium, inconel, waspaloy, nimonic, and cast iron. It can be run at speeds up to 400 SFM. The end mill is made in Japan by Kyocera and is sold individually.