Learn More
Grainger End Mill Kyocera MFPN 452000RW1253T 2.
Catalog No. 502764672
Encompass_Preferred
The KYOCERA Indexable Face Mill 2-3100-170HK6 is a face milling cutter with a 2.5" diameter and a maximum depth of cut of 0.5". It has 6 indexable carbide inserts with the 170HK6 grade for high speed steel and hardened materials. The cutter has a right-hand spiral flute design and accepts inserts with a screw clamping system. It is balanced for high RPM and has a 1" arbor hole with a keyway drive. The overall length is 3.1" and it weighs 1.1 lbs. This durable and versatile face mill is ideal for heavy duty facing, slotting and profiling applications.
Catalog No. 50-276-4672 Supplier Grainger Supplier No. 170HK6
The KYOCERA Indexable Face Mill 2-3100-170HK6 is a face milling cutter with a 2.5" diameter and a maximum depth of cut of 0.5". It has 6 indexable carbide inserts with the 170HK6 grade for high speed steel and hardened materials. The cutter has a right-hand spiral flute design and accepts inserts with a screw clamping system. It is balanced for high RPM and has a 1" arbor hole with a keyway drive. The overall length is 3.1" and it weighs 1.1 lbs. This durable and versatile face mill is ideal for heavy duty facing, slotting and profiling applications.