Learn More
Grainger End Mill Kyocera MFWN 90050RS323T 50.0
Catalog No. 502763836
Encompass_Preferred
The KYOCERA Indexable Face Mill 50.00mm 170HR9 is a high-performance face milling cutter with a 50mm diameter. It features a 170HR9 grade carbide insert with a positive rake angle for smooth milling in a wide range of materials including steel, stainless steel, cast iron, and hardened materials up to 45HRC. The cutter body is made of alloy steel for durability. It has Weldon style flat shanks and accepts SEHT1204AFN inserts with four usable cutting edges. This versatile face mill is well-suited for light to heavy milling applications.
Catalog No. 50-276-3836 Supplier Grainger Supplier No. 170HR9
The KYOCERA Indexable Face Mill 50.00mm 170HR9 is a high-performance face milling cutter with a 50mm diameter. It features a 170HR9 grade carbide insert with a positive rake angle for smooth milling in a wide range of materials including steel, stainless steel, cast iron, and hardened materials up to 45HRC. The cutter body is made of alloy steel for durability. It has Weldon style flat shanks and accepts SEHT1204AFN inserts with four usable cutting edges. This versatile face mill is well-suited for light to heavy milling applications.