Learn More
Charles River Laboratories Endpoint Chromogenic Kit, 140 tests
Supplier:  Charles River Laboratories R160K
Encompass
5 × 1.4 mL vials of chromogenic LAL, 1 × 10 mg vial of chromogenic substrate S-2423, 2 × 2 ng vials of endotoxin, 2 × 30 mL vials of LAL reagent water, and 1 × 15 mL 0.05 M vial of Tris buffer.
Catalog No. NC9593644