Learn More
McMaster-Carr Terminal Block End Stops
Catalog No. NC1457116
Encompass
The McMaster-Carr terminal block end stops are used to prevent overtravel on sliding mechanisms. They have a low profile design that mounts directly to the end of a T-slotted rail. The end stops are available in black thermoplastic or stainless steel construction. The thermoplastic version can withstand temperatures up to 200°F continuous. The stainless steel version is rated for temperatures up to 1000°F continuous. Both versions feature a spring-loaded plunger to absorb impact. The end stops come in a variety of sizes to accommodate different profile rail widths. Overall, these end stops provide a durable and customizable solution for limiting travel on linear motion systems.
Catalog No. NC1457116 Supplier McMaster-Carr Supplier No. 7641K73
The McMaster-Carr terminal block end stops are used to prevent overtravel on sliding mechanisms. They have a low profile design that mounts directly to the end of a T-slotted rail. The end stops are available in black thermoplastic or stainless steel construction. The thermoplastic version can withstand temperatures up to 200°F continuous. The stainless steel version is rated for temperatures up to 1000°F continuous. Both versions feature a spring-loaded plunger to absorb impact. The end stops come in a variety of sizes to accommodate different profile rail widths. Overall, these end stops provide a durable and customizable solution for limiting travel on linear motion systems.