Learn More
Pyramex™ Endeavor Plus Anti-Fog Safety Glasses
Catalog No. 19555701 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Frame Color:
Black, Gray
Black, Green
Black, Pink
Lens Coating:
Anti-fog
Anti-fog, Anti-scratch
Lens Tint:
Clear
Gray
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Coating Lens Tint
19-555-701 Black, Pink Anti-fog Clear
19-555-691 Black, Gray Anti-fog, Anti-scratch Clear
19-555-692 Black, Gray Anti-fog, Anti-scratch Gray
19-555-694 Black, Green Anti-fog, Anti-scratch Clear
19-555-695 Black, Green Anti-fog, Anti-scratch Gray
19-555-693 Black, Green Anti-fog Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-555-701 Supplier Pyramex™ Supplier No. SBP9610ST
Add to Cart
Add to Cart
This item is not returnable. View return policy
Endeavor Plus Anti-Fog Safety Glasses with ANSI Z87 safety compliance
  • Lightweight and durable protective eyewear
  • Built-in brow guard shields against overhead particles
  • Special dielectric hinge design contains no metal parts
  • Curved, co-material temples allow for comfortable all day wear
  • Scratch resistant polycarbonate lens
  • 9.5 base one-piece wraparound lens provides excellent side protection
  • Soft and adjustable nose piece provides a customized fit to prevent slipping
  • Lens Technology:
    • Visible Light Transmitted - 23%
    • UVA, UVB Blocked - 100%
    • Blue Light Blocked - 74%
    • IR Blocked - 48%
Specifications
Lens Coating Anti-fog
Lens Type H2X
Certifications/Compliance ANSI Z87
Lens Tint Clear
For Use With (Application) Eye Safety
Frame Color Black, Pink
Temple Style Curved Co-Injected
Lens Material Polycarbonate
Frame Material Polycarbonate, TPE
UV Protection UV400