Learn More
Pyramex™ Endeavor™ Plus Safety Eyewear
Catalog No. 19155910 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Coating:
Anti-fog, Scratch-resistant
Scratch-resistant
Frame Color:
Black, Gray
Black, Green
Black, Pink
Lens Tint:
Clear
Gray
Indoor/Outdoor, Mirror
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating Frame Color Lens Tint
19-155-910 Scratch-resistant Black, Pink Clear
19-155-900 Scratch-resistant Black, Gray Clear
19-155-901 Anti-fog, Scratch-resistant Black, Gray Clear
19-155-902 Scratch-resistant Black, Gray Gray
19-155-903 Anti-fog, Scratch-resistant Black, Gray Gray
19-155-905 Scratch-resistant Black, Gray Indoor/Outdoor, Mirror
19-155-906 Scratch-resistant Black, Green Clear
19-155-907 Scratch-resistant Black, Green Gray
19-155-908 Scratch-resistant Black, Green Indoor/Outdoor, Mirror
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 19-155-910 Supplier Pyramex™ Supplier No. SBP9610S
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Lightweight and durable protective eyewear
  • Built-in brow guard shields against overhead particles
  • Special dielectric hinge design contains no metal parts
  • Curved, co-material temples allow for comfortable all day wear
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • 9.5 base one-piece wraparound lens provides excellent side protection
  • Soft and adjustable nose piece provides a customized fit to prevent slippping
  • Specifications
    Lens Coating Scratch-resistant
    Certifications/Compliance ANSI Z87.1,CSA Z94.3-07
    Lens Tint Clear
    Frame Color Black, Pink
    Weight (Metric) 23.5 g
    Lens Material Polycarbonate
    Frame Material Polycarbonate, TPE