Learn More
Tosoh Bioscience LLC ENDFITTING 10UM FRIT FOR 5MMGL
Supplier:  Tosoh Bioscience LLC 0016104
Encompass
Fitting, End; With 10 micrometer Pyrex frit for 5mm I.D. glass column; Tosoh
Catalog No. 50-851-389