Learn More
Broadpharm endo-BCN CE-phosphoramidite, 500 mg, Copper-Free Click Chemistry, Azides (SPAAC), Tetrazines (IEDDA)
Catalog No. NC2875171
Encompass
endo-BCN CE-phosphoramidite is a highly reactive bicyclononyne (BCN) derivative designed for efficient oligonucleotide synthesis and bioconjugation via copper-free click chemistry. This stable cyclooctyne allows for versatile reactivity with azides and tetrazines, enhancing cycloaddition rates. The *endo*-stereoisomer makes it effective for incorporating BCN labels into oligonucleotides, facilitating conjugation to a variety of substrates. It is compatible with standard oligonucleotide synthesis protocols, enhancing ease of use in diverse applications.
  • Copper-free click chemistry facilitates bioorthogonal conjugations.
  • Reacts efficiently with both azides and tetrazines.
  • Endo-stereoisomer configuration promotes higher cycloaddition rates.
  • Enables incorporation of BCN labels for broader labeling strategies.
  • Usable with standard synthesis conditions.
Catalog No. NC2875171 Supplier Broadpharm Supplier No. BP40265500MG
endo-BCN CE-phosphoramidite is a highly reactive bicyclononyne (BCN) derivative designed for efficient oligonucleotide synthesis and bioconjugation via copper-free click chemistry. This stable cyclooctyne allows for versatile reactivity with azides and tetrazines, enhancing cycloaddition rates. The *endo*-stereoisomer makes it effective for incorporating BCN labels into oligonucleotides, facilitating conjugation to a variety of substrates. It is compatible with standard oligonucleotide synthesis protocols, enhancing ease of use in diverse applications.
  • Copper-free click chemistry facilitates bioorthogonal conjugations.
  • Reacts efficiently with both azides and tetrazines.
  • Endo-stereoisomer configuration promotes higher cycloaddition rates.
  • Enables incorporation of BCN labels for broader labeling strategies.
  • Usable with standard synthesis conditions.