Learn More
Lumiprobe ENDO-BCN CE-Phosphoramidite, 1 g, C24H40N3O5P, 481.57 g/mol, reactive with azides and tetrazines
Catalog No. NC3404311
Encompass_Preferred
ENDO-BCN CE-Phosphoramidite, 1 g, C24H40N3O5P, 481.57 g/mol, reactive with azides and tetrazines
Catalog No. NC3404311 Supplier Lumiprobe Supplier No. 30051G
ENDO-BCN CE-Phosphoramidite, 1 g, C24H40N3O5P, 481.57 g/mol, reactive with azides and tetrazines