Learn More
Medchemexpress LLC Endo-BCN-NHS carbonate | 1426827-79-3 | MFCD19705416 | ≥97.0% | 291.30 g·mol⁻1 | C15H17NO5 | 500 MG
SDP
Catalog No. 5000299842
Encompass_Preferred
endo-BCN-NHS carbonate is an NHS-activated bicyclo[6.1.0]non-4-yne (BCN) carbonate reagent for bioconjugation. It couples to primary amines to install a strained cyclooctyne handle for strain-promoted azide-alkyne cycloaddition (SPAAC), enabling copper-free bioorthogonal labeling of proteins, peptides, and polymers.
  • NHS-activated carbonate enables efficient coupling to primary amines.
  • Introduces a strained BCN handle for copper-free SPAAC reactions.
  • High purity suitable for analytical and preparative bioconjugation.
  • Solid form with recommended storage at -20°C for prolonged stability.
  • Compatible with proteins, peptides, and polymer modification workflows.
Catalog No. 50-002-99842 Supplier Medchemexpress LLC Supplier No. HYW035129500MG
endo-BCN-NHS carbonate is an NHS-activated bicyclo[6.1.0]non-4-yne (BCN) carbonate reagent for bioconjugation. It couples to primary amines to install a strained cyclooctyne handle for strain-promoted azide-alkyne cycloaddition (SPAAC), enabling copper-free bioorthogonal labeling of proteins, peptides, and polymers.
  • NHS-activated carbonate enables efficient coupling to primary amines.
  • Introduces a strained BCN handle for copper-free SPAAC reactions.
  • High purity suitable for analytical and preparative bioconjugation.
  • Solid form with recommended storage at -20°C for prolonged stability.
  • Compatible with proteins, peptides, and polymer modification workflows.