Learn More
Medchemexpress LLC Endo-BCN-NHS carbonate | 1426827-79-3 | MFCD19705416 | 99.5% | 291.30 g·mol⁻¹ | C15H17NO5 | 50 MG
SDP
Catalog No. 5000299535
Encompass_Preferred
endo-BCN-NHS carbonate is an NHS-activated bicyclononyne carbonate reagent used for bioconjugation. It functionalizes primary amines to introduce BCN handles that enable strain-promoted azide-alkyne cycloaddition (SPAAC) for copper-free click chemistry, commonly applied to modify elastin-like proteins and to form cross-linked hydrogels for cell encapsulation studies.
  • Reacts with primary amines to install BCN functional groups.
  • Enables copper-free strain-promoted azide-alkyne cycloaddition (SPAAC).
  • Facilitates hydrogel formation and protein cross-linking for cell encapsulation.
  • High purity supports reproducible conjugation chemistry.
  • Powder stable at -20°C for long-term storage; solvent stability varies.
Catalog No. 50-002-99535 Supplier Medchemexpress LLC Supplier No. HYW03512950MG
endo-BCN-NHS carbonate is an NHS-activated bicyclononyne carbonate reagent used for bioconjugation. It functionalizes primary amines to introduce BCN handles that enable strain-promoted azide-alkyne cycloaddition (SPAAC) for copper-free click chemistry, commonly applied to modify elastin-like proteins and to form cross-linked hydrogels for cell encapsulation studies.
  • Reacts with primary amines to install BCN functional groups.
  • Enables copper-free strain-promoted azide-alkyne cycloaddition (SPAAC).
  • Facilitates hydrogel formation and protein cross-linking for cell encapsulation.
  • High purity supports reproducible conjugation chemistry.
  • Powder stable at -20°C for long-term storage; solvent stability varies.