Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Endo-BCN-PEG2-alcohol | 1807501-85-4 | 98.0% | 325.40 | 100 MG
SDP

Catalog No. 5000349856
Encompass_Preferred

endo-BCN-PEG2-alcohol is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring a BCN group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups. PROTACs consist of two different ligands connected by a linker; one for an E3 ubiquitin ligase and the other for the target protein. PROTACs degrade target proteins using the intracellular ubiquitin-proteasome system.

  • Purity: 98.0%
  • Molecular weight: 325.40
  • Formula: C17H27NO5
  • Appearance: Oil
  • Color: Colorless to light yellow
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: -20°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light)

Catalog No. 50-003-49856 Supplier Medchemexpress LLC Supplier No. HY140076100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

endo-BCN-PEG2-alcohol is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring a BCN group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups. PROTACs consist of two different ligands connected by a linker; one for an E3 ubiquitin ligase and the other for the target protein. PROTACs degrade target proteins using the intracellular ubiquitin-proteasome system.

  • Purity: 98.0%
  • Molecular weight: 325.40
  • Formula: C17H27NO5
  • Appearance: Oil
  • Color: Colorless to light yellow
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: -20°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.