Learn More
Medchemexpress LLC endo-BCN-PEG2-alcohol | 1807501-85-4 | 98.0% | 325.40 | 25 MG
SDP
Catalog No. 5000349413
Encompass_Preferred
endo-BCN-PEG2-alcohol is a PEG-based PROTAC linker employed in the synthesis of PROTACs. This compound functions as a click chemistry reagent, featuring a BCN group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups.
  • PEG-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • Functions as a click chemistry reagent.
  • Contains a BCN group for strain-promoted alkyne-azide cycloaddition (SPAAC).
Catalog No. 50-003-49413 Supplier Medchemexpress LLC Supplier No. 25MGHY140076
endo-BCN-PEG2-alcohol is a PEG-based PROTAC linker employed in the synthesis of PROTACs. This compound functions as a click chemistry reagent, featuring a BCN group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups.
  • PEG-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • Functions as a click chemistry reagent.
  • Contains a BCN group for strain-promoted alkyne-azide cycloaddition (SPAAC).