Learn More
Medchemexpress LLC Endo-BCN-PEG2-alcohol | 1807501-85-4 | 98.0% | 325.40 | 50 MG
SDP
Catalog No. 5000349202
Encompass_Preferred
Endo-BCN-PEG2-alcohol is a versatile PEG-based PROTAC linker. It plays a crucial role in the synthesis of PROTACs by facilitating the connection between target proteins and E3 ubiquitin ligases. Additionally, it functions as a click chemistry reagent, enabling efficient and specific bioconjugation reactions.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains a BCN group for strain-promoted alkyne-azide cycloaddition (SPAAC) with azide groups
Catalog No. 50-003-49202 Supplier Medchemexpress LLC Supplier No. HY14007650MG
Endo-BCN-PEG2-alcohol is a versatile PEG-based PROTAC linker. It plays a crucial role in the synthesis of PROTACs by facilitating the connection between target proteins and E3 ubiquitin ligases. Additionally, it functions as a click chemistry reagent, enabling efficient and specific bioconjugation reactions.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains a BCN group for strain-promoted alkyne-azide cycloaddition (SPAAC) with azide groups