Learn More
Medchemexpress LLC Endo-BCN-PEG2-alcohol | 1807501-85-4 | 98.0% | C17H27NO5 | 10MG
SDP
Catalog No. 5000260401
Encompass_Preferred
endo-BCN-PEG2-alcohol is a PEG-based linker that combines a strained bicyclo[6.1.0]non-4-yne (BCN) reactive group with a terminal hydroxyl. It is intended for use in copper-free click chemistry, bioconjugation, and linker assembly in PROTAC synthesis.
  • Contains BCN strained alkyne for copper-free click reactions.
  • Terminal alcohol enables further functionalization.
  • PEG2 spacer increases hydrophilicity and reduces steric hindrance.
  • Suitable for PROTAC linker assembly and bioconjugation applications.
  • Comes with analytical documentation and storage recommendations.
Catalog No. 50-002-60401 Supplier Medchemexpress LLC Supplier No. HY14007610MG
endo-BCN-PEG2-alcohol is a PEG-based linker that combines a strained bicyclo[6.1.0]non-4-yne (BCN) reactive group with a terminal hydroxyl. It is intended for use in copper-free click chemistry, bioconjugation, and linker assembly in PROTAC synthesis.
  • Contains BCN strained alkyne for copper-free click reactions.
  • Terminal alcohol enables further functionalization.
  • PEG2 spacer increases hydrophilicity and reduces steric hindrance.
  • Suitable for PROTAC linker assembly and bioconjugation applications.
  • Comes with analytical documentation and storage recommendations.