Learn More
Medchemexpress LLC Endo-BCN-PEG2-C2-NHS ester | 2243565-12-8 | 95.0% | 450.48 | 100 MG
SDP
Catalog No. 5000349765
Encompass_Preferred
endo-BCN-PEG2-C2-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets a protein of interest. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-49765 Supplier Medchemexpress LLC Supplier No. HY140067100MG
endo-BCN-PEG2-C2-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets a protein of interest. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins