Learn More
Medchemexpress LLC Endo-BCN-PEG2-C2-NHS ester | 2243565-12-8 | 95.0% | 450.48 | 25 MG
SDP
Catalog No. 5000348735
Encompass_Preferred
Endo-BCN-PEG2-C2-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Facilitates the connection of two different ligands (E3 ubiquitin ligase ligand and target protein ligand)
  • Exploits the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins
Catalog No. 50-003-48735 Supplier Medchemexpress LLC Supplier No. 25MGHY140067
Endo-BCN-PEG2-C2-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Facilitates the connection of two different ligands (E3 ubiquitin ligase ligand and target protein ligand)
  • Exploits the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins