Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Endo-BCN-PEG2-C2-NHS ester | 2243565-12-8 | 95.0% | 450.48 | 50 MG
SDP

Catalog No. 5000349851
Encompass_Preferred

Endo-BCN-PEG2-C2-NHS ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These compounds consist of two distinct ligands connected by a linker: one ligand targets an E3 ubiquitin ligase, and the other targets a specific protein. PROTACs operate by harnessing the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Composed of two different ligands connected by a linker
  • Targets an E3 ubiquitin ligase and a target protein
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
  • Molecular weight: 450.48
  • Appearance: Oil
  • Color: Light yellow to yellow

Catalog No. 50-003-49851 Supplier Medchemexpress LLC Supplier No. HY14006750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Endo-BCN-PEG2-C2-NHS ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These compounds consist of two distinct ligands connected by a linker: one ligand targets an E3 ubiquitin ligase, and the other targets a specific protein. PROTACs operate by harnessing the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Composed of two different ligands connected by a linker
  • Targets an E3 ubiquitin ligase and a target protein
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
  • Molecular weight: 450.48
  • Appearance: Oil
  • Color: Light yellow to yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.