Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Endo-BCN-PEG2-C2-NHS ester | 2243565-12-8 | 95.0% | C22H30N2O8 | 10MG
SDP

Catalog No. 5000260193
Encompass_Preferred

endo-BCN-PEG2-C2-NHS ester is a PEG-based bifunctional linker for PROTAC construction and bioconjugation. It combines a strained endo-bicyclononyne (BCN) for copper-free click chemistry with an N-hydroxysuccinimide (NHS) ester for selective amine coupling, enabling efficient assembly of bifunctional molecules and protein labeling.

  • Contains endo-bicyclononyne (BCN) for strain-promoted azide-alkyne cycloaddition
  • Includes an N-hydroxysuccinimide (NHS) ester for rapid amine conjugation
  • Incorporates a PEG2 spacer to improve solubility and reduce steric hindrance
  • Suitable for PROTAC linker synthesis and bioconjugation workflows
  • Molecular formula C22H30N2O8 and molecular weight 450.48 g/mol

Catalog No. 50-002-60193 Supplier Medchemexpress LLC Supplier No. HY14006710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

endo-BCN-PEG2-C2-NHS ester is a PEG-based bifunctional linker for PROTAC construction and bioconjugation. It combines a strained endo-bicyclononyne (BCN) for copper-free click chemistry with an N-hydroxysuccinimide (NHS) ester for selective amine coupling, enabling efficient assembly of bifunctional molecules and protein labeling.

  • Contains endo-bicyclononyne (BCN) for strain-promoted azide-alkyne cycloaddition
  • Includes an N-hydroxysuccinimide (NHS) ester for rapid amine conjugation
  • Incorporates a PEG2 spacer to improve solubility and reduce steric hindrance
  • Suitable for PROTAC linker synthesis and bioconjugation workflows
  • Molecular formula C22H30N2O8 and molecular weight 450.48 g/mol

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.