Learn More
Medchemexpress LLC Endo-BCN-PEG2-C2-NHS ester | 2243565-12-8 | MFCD19705420 | 95.0% | 450.48 g/mol | C22H30N2O8 | 5 MG
SDP
Catalog No. 5000222563
Encompass_Preferred
endo-BCN-PEG2-C2-NHS ester is a BCN-containing PEG-based N-hydroxysuccinimide (NHS) ester linker for bioconjugation and PROTAC synthesis. The NHS ester enables efficient coupling to primary amines, while the BCN moiety provides a strained alkyne handle for copper-free click reactions, supporting linker installation in small molecules and selective protein labeling workflows.
  • Reacts with primary amines via NHS ester chemistry.
  • Provides strained alkyne (BCN) for copper-free click chemistry.
  • Suitable for PROTAC linker installation and protein labeling.
  • High purity for consistent coupling performance.
  • Available in small-scale packages for research use.
Catalog No. 50-002-22563 Supplier Medchemexpress LLC Supplier No. HY1400675MG
endo-BCN-PEG2-C2-NHS ester is a BCN-containing PEG-based N-hydroxysuccinimide (NHS) ester linker for bioconjugation and PROTAC synthesis. The NHS ester enables efficient coupling to primary amines, while the BCN moiety provides a strained alkyne handle for copper-free click reactions, supporting linker installation in small molecules and selective protein labeling workflows.
  • Reacts with primary amines via NHS ester chemistry.
  • Provides strained alkyne (BCN) for copper-free click chemistry.
  • Suitable for PROTAC linker installation and protein labeling.
  • High purity for consistent coupling performance.
  • Available in small-scale packages for research use.