Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Endo-BCN-PEG2-NH2 | 1263166-93-3 | 97.0% | 324.42 | C17H28N2O4 | 50 MG
SDP

Catalog No. 5000210830
Encompass_Preferred

Endo-BCN-PEG2-NH2 is a PEG-based linker bearing a strained bicyclo[6.1.0]non-4-yne (BCN) alkyne and a terminal primary amine. It is designed for strain-promoted alkyne-azide cycloaddition (SPAAC) click chemistry and is used in PROTAC synthesis and bioconjugation workflows.

  • Presents BCN strained alkyne for SPAAC conjugation.
  • Contains a PEG2 spacer for improved solubility and flexibility.
  • Terminal primary amine available for further functionalization.
  • High purity (NMR): 97.0%.
  • Molecular weight: 324.42 g/mol; formula C17H28N2O4.

Catalog No. 50-002-10830 Supplier Medchemexpress LLC Supplier No. HYW07275250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Endo-BCN-PEG2-NH2 is a PEG-based linker bearing a strained bicyclo[6.1.0]non-4-yne (BCN) alkyne and a terminal primary amine. It is designed for strain-promoted alkyne-azide cycloaddition (SPAAC) click chemistry and is used in PROTAC synthesis and bioconjugation workflows.

  • Presents BCN strained alkyne for SPAAC conjugation.
  • Contains a PEG2 spacer for improved solubility and flexibility.
  • Terminal primary amine available for further functionalization.
  • High purity (NMR): 97.0%.
  • Molecular weight: 324.42 g/mol; formula C17H28N2O4.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.