Learn More
Medchemexpress LLC Endo-BCN-PEG3-maleimide | 2141976-33-0 | MFCD31811466 | 98.5% | 519.59 g/mol | C26H37N3O8 | 5 MG
SDP
Catalog No. 5000262529
Encompass_Preferred
endo-BCN-PEG3-maleimide is a heterobifunctional PEG3 linker that pairs an endo-bicyclononyne (BCN) for strain-promoted azide-alkyne cycloaddition with a maleimide for thiol-selective conjugation. The short PEG3 spacer improves solubility and flexibility, making the reagent suitable for bioconjugation, PROTAC linker assembly, and crosslinking in research applications. Supplied as a colorless to light yellow oil with high reported purity and recommended cold storage.
  • Strain-promoted azide-alkyne cycloaddition reactivity (BCN).
  • Maleimide-based thiol conjugation for cysteine targeting.
  • PEG3 spacer improves solubility and flexibility.
  • High purity suitable for research use.
  • Stable under low-temperature storage conditions.
  • Soluble in common organic solvents such as DCM, THF, ACN, DMF, and DMSO.
Catalog No. 50-002-62529 Supplier Medchemexpress LLC Supplier No. HY1334005MG
endo-BCN-PEG3-maleimide is a heterobifunctional PEG3 linker that pairs an endo-bicyclononyne (BCN) for strain-promoted azide-alkyne cycloaddition with a maleimide for thiol-selective conjugation. The short PEG3 spacer improves solubility and flexibility, making the reagent suitable for bioconjugation, PROTAC linker assembly, and crosslinking in research applications. Supplied as a colorless to light yellow oil with high reported purity and recommended cold storage.
  • Strain-promoted azide-alkyne cycloaddition reactivity (BCN).
  • Maleimide-based thiol conjugation for cysteine targeting.
  • PEG3 spacer improves solubility and flexibility.
  • High purity suitable for research use.
  • Stable under low-temperature storage conditions.
  • Soluble in common organic solvents such as DCM, THF, ACN, DMF, and DMSO.