Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Endo-BCN-PEG4-acid | 1881221-47-1 | 99.3% | 441.52 | C22H35NO8 | 10 MG
SDP

Catalog No. 5000203680
Encompass_Preferred

endo-BCN-PEG4-acid is a cleavable four-unit polyethylene glycol (PEG) linker containing a bicyclononyne (BCN) group that enables strain-promoted alkyne-azide cycloaddition (SPAAC). It is used as a copper-free click chemistry reagent for bioconjugation, including antibody-drug conjugate and PROTAC linker synthesis.

  • Cleavable 4-unit PEG linker facilitating conjugation.
  • BCN moiety enables copper-free SPAAC with azide groups.
  • Molecular weight 441.52 and formula C22H35NO8.
  • CAS number 1881221-47-1; high purity for research use.
  • Storage recommendations include low-temperature conditions for long-term stability.
  • Typical applications: ADC linker chemistry, bioconjugation, and PROTAC synthesis.

Catalog No. 50-002-03680 Supplier Medchemexpress LLC Supplier No. HY13597110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

endo-BCN-PEG4-acid is a cleavable four-unit polyethylene glycol (PEG) linker containing a bicyclononyne (BCN) group that enables strain-promoted alkyne-azide cycloaddition (SPAAC). It is used as a copper-free click chemistry reagent for bioconjugation, including antibody-drug conjugate and PROTAC linker synthesis.

  • Cleavable 4-unit PEG linker facilitating conjugation.
  • BCN moiety enables copper-free SPAAC with azide groups.
  • Molecular weight 441.52 and formula C22H35NO8.
  • CAS number 1881221-47-1; high purity for research use.
  • Storage recommendations include low-temperature conditions for long-term stability.
  • Typical applications: ADC linker chemistry, bioconjugation, and PROTAC synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.