Learn More
Medchemexpress LLC Endo-BCN-PEG4-acid | 1881221-47-1 | 99.3% | 441.52 g/mol | C22H35NO8 | 100 MG
SDP
Catalog No. 5000202583
Encompass_Preferred
endo-BCN-PEG4-acid is a BCN-containing PEG4 linker used for click chemistry and bioconjugation, typically employed in the synthesis of antibody-drug conjugates (ADCs) via strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Cleavable four-unit PEG linker suitable for ADC and PROTAC synthesis.
  • Contains a strained BCN moiety for efficient SPAAC with azide-containing partners.
  • High purity for conjugation applications.
  • Liquid form, colorless to light yellow, with recommended low-temperature storage.
  • Available as a 100 MG pack for laboratory use.
Catalog No. 50-002-02583 Supplier Medchemexpress LLC Supplier No. HY135971100MG
endo-BCN-PEG4-acid is a BCN-containing PEG4 linker used for click chemistry and bioconjugation, typically employed in the synthesis of antibody-drug conjugates (ADCs) via strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Cleavable four-unit PEG linker suitable for ADC and PROTAC synthesis.
  • Contains a strained BCN moiety for efficient SPAAC with azide-containing partners.
  • High purity for conjugation applications.
  • Liquid form, colorless to light yellow, with recommended low-temperature storage.
  • Available as a 100 MG pack for laboratory use.