Learn More
Medchemexpress LLC Endo-BCN-PEG4-acid | 1881221-47-1 | 99.3% | 441.52 g/mol | C22H35NO8 | 5 MG
SDP
Catalog No. 5000203715
Encompass_Preferred
endo-BCN-PEG4-acid is a BCN-containing PEG4 linker used for bioconjugation and ADC synthesis. It enables strain-promoted alkyne-azide cycloaddition (SPAAC) click chemistry and provides a terminal carboxylic acid for covalent coupling. Supplied as a high-purity liquid for research applications.
  • Cleavable PEG4 linker for bioconjugation.
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Terminal carboxylic acid for amide coupling.
  • High reported purity (99.3%).
  • Liquid, colorless to light yellow; store frozen for long-term stability.
  • For research use only; not for human therapeutic use.
Catalog No. 50-002-03715 Supplier Medchemexpress LLC Supplier No. HY1359715MG
endo-BCN-PEG4-acid is a BCN-containing PEG4 linker used for bioconjugation and ADC synthesis. It enables strain-promoted alkyne-azide cycloaddition (SPAAC) click chemistry and provides a terminal carboxylic acid for covalent coupling. Supplied as a high-purity liquid for research applications.
  • Cleavable PEG4 linker for bioconjugation.
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Terminal carboxylic acid for amide coupling.
  • High reported purity (99.3%).
  • Liquid, colorless to light yellow; store frozen for long-term stability.
  • For research use only; not for human therapeutic use.