Learn More
Broadpharm endo-BCN-PEG4-NHS ester | 1807501-86-5 | MFCD23704191 | 100mg
Catalog No. NC1542846
Encompass
endo-BCN-PEG4-NHS ester | Purity: 95% | MW: 538.59 | 1807501-86-5 | MFCD23704191 | 100mg
Catalog No. NC1542846 Supplier Broadpharm Supplier No. BP22851100MG
endo-BCN-PEG4-NHS ester | Purity: 95% | MW: 538.59 | 1807501-86-5 | MFCD23704191 | 100mg