Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Broadpharm endo-BCN-PEG4-NHS ester, 500 mg, C26H38N2O10, molecular weight 538.6 g/mol, NHS ester, BCN, DMSO/DMF/DCM soluble, amine-reactive, azide-reactive

Catalog No. NC3444739
Encompass

The endo-BCN-PEG4-NHS ester is a versatile reagent specifically designed for bioconjugation applications, facilitating the labeling of biomolecules such as proteins, peptides, and oligonucleotides. With its unique functional groups, this heterobifunctional PEG derivative offers a reliable method for biomolecule modification through efficient click chemistry. Ideal for research in PROTAC synthesis and various advanced bioconjugation techniques, this product enhances solubility in aqueous environments, ensuring optimal performance in scientific applications.

  • NHS ester group enables easy labeling of primary amines.
  • BCN moiety allows for rapid, copper-free click reactions.
  • PEG4 spacer increases solubility in aqueous media.
  • Reagent grade, suitable for research purposes.
  • Available quantity: 500 mg.
  • Storage conditions at -20°C for stability.
  • Applications include PROTAC synthesis and bioconjugation.

Catalog No. NC3444739 Supplier Broadpharm Supplier No. BP22851500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The endo-BCN-PEG4-NHS ester is a versatile reagent specifically designed for bioconjugation applications, facilitating the labeling of biomolecules such as proteins, peptides, and oligonucleotides. With its unique functional groups, this heterobifunctional PEG derivative offers a reliable method for biomolecule modification through efficient click chemistry. Ideal for research in PROTAC synthesis and various advanced bioconjugation techniques, this product enhances solubility in aqueous environments, ensuring optimal performance in scientific applications.

  • NHS ester group enables easy labeling of primary amines.
  • BCN moiety allows for rapid, copper-free click reactions.
  • PEG4 spacer increases solubility in aqueous media.
  • Reagent grade, suitable for research purposes.
  • Available quantity: 500 mg.
  • Storage conditions at -20°C for stability.
  • Applications include PROTAC synthesis and bioconjugation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.