Learn More
Medchemexpress LLC endo-BCN-PEG4-PFP ester | 2904607-26-5 | 95.0% | 607.56 | 100 MG
SDP
Catalog No. 5000349480
Encompass_Preferred
endo-BCN-PEG4-PFP ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent, featuring a BCN group capable of undergoing strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups.
  • PEG-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • Click chemistry reagent.
  • Contains a BCN group that undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) with azide groups.
  • PROTACs utilize the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
Catalog No. 50-003-49480 Supplier Medchemexpress LLC Supplier No. HY140073100MG
endo-BCN-PEG4-PFP ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent, featuring a BCN group capable of undergoing strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups.
  • PEG-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • Click chemistry reagent.
  • Contains a BCN group that undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) with azide groups.
  • PROTACs utilize the intracellular ubiquitin-proteasome system for selective degradation of target proteins.