Learn More
Medchemexpress LLC Endo-BCN-PEG4-PFP ester | 2904607-26-5 | 95.0% | C28H34F5NO8 | 10MG
SDP
Catalog No. 5000262029
Encompass_Preferred
endo-BCN-PEG4-PFP ester is a PEG-based bifunctional linker combining a strained alkyne (BCN) for strain-promoted alkyne-azide cycloaddition with a pentafluorophenyl (PFP) activated ester for efficient conjugation to primary amines. Supplied as an oil, it is designed for use in click chemistry, bioconjugation, and linker assembly in targeted degrader synthesis.
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) without copper.
  • PFP ester reacts efficiently with primary amines to form stable amide bonds.
  • PEG4 spacer improves solubility and reduces steric hindrance.
  • Suitable for bioconjugation and targeted degrader linker construction workflows.
  • Stable when stored frozen; follow recommended storage conditions for longevity.
Catalog No. 50-002-62029 Supplier Medchemexpress LLC Supplier No. HY14007310MG
endo-BCN-PEG4-PFP ester is a PEG-based bifunctional linker combining a strained alkyne (BCN) for strain-promoted alkyne-azide cycloaddition with a pentafluorophenyl (PFP) activated ester for efficient conjugation to primary amines. Supplied as an oil, it is designed for use in click chemistry, bioconjugation, and linker assembly in targeted degrader synthesis.
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) without copper.
  • PFP ester reacts efficiently with primary amines to form stable amide bonds.
  • PEG4 spacer improves solubility and reduces steric hindrance.
  • Suitable for bioconjugation and targeted degrader linker construction workflows.
  • Stable when stored frozen; follow recommended storage conditions for longevity.