Learn More
Medchemexpress LLC Endo-BCN-PEG8-NHS ester | 1608140-48-2 | 95.0% | 714.80 | 50 MG
SDP
Catalog No. 5000349771
Encompass_Preferred
Endo-BCN-PEG8-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring a BCN group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules. This product is utilized in leveraging the intracellular ubiquitin-proteasome system for selective protein degradation.
  • PEG-based PROTAC linker
  • Functions as a click chemistry reagent
  • Contains a BCN group for strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Facilitates synthesis of PROTACs
  • Supports targeted protein degradation
Catalog No. 50-003-49771 Supplier Medchemexpress LLC Supplier No. HY14007050MG
Endo-BCN-PEG8-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring a BCN group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules. This product is utilized in leveraging the intracellular ubiquitin-proteasome system for selective protein degradation.
  • PEG-based PROTAC linker
  • Functions as a click chemistry reagent
  • Contains a BCN group for strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Facilitates synthesis of PROTACs
  • Supports targeted protein degradation