Learn More
Medchemexpress LLC endo-BCN-PEG8-NHS ester, 25mg | CAS No. : 1608140-48-2
SDP
Catalog No. NC3787632
Encompass_Preferred
endo-BCN-PEG8-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. endo-BCN-PEG8-NHS ester is a click chemistry reagent, it contains a BCN group that can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups.
Catalog No. NC3787632 Supplier Medchemexpress LLC Supplier No. HY14007025MG
endo-BCN-PEG8-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. endo-BCN-PEG8-NHS ester is a click chemistry reagent, it contains a BCN group that can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups.