Learn More
Medchemexpress LLC endo CNTinh-03 | 345288-49-5 | 100.0% | 427.51 | 1 ML
SDP
Catalog No. 5000401573
Encompass_Preferred
Inhibitor for the elevation of cAMP and cGMP induced by agonist, such as G protein-coupled receptors, adenylate cyclase, and guanylate cyclase (IC50 is 4 μM). It inhibits cholera toxin- and Escherichia coli (STa) toxin- induced CFTR chloride current, ameliorates secretory diarrhea in mouse models, and prevents cyst growth in polycystic kidney disease model.
  • Inhibitor for the elevation of cAMP and cGMP induced by agonist.
  • Inhibits cholera toxin- and Escherichia coli (STa) toxin- induced CFTR chloride current.
  • Ameliorates secretory diarrhea in mouse models.
  • Prevents cyst growth in polycystic kidney disease model.
Catalog No. 50-004-01573 Supplier Medchemexpress LLC Supplier No. HY40191310MM/1ML
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Inhibitor for the elevation of cAMP and cGMP induced by agonist, such as G protein-coupled receptors, adenylate cyclase, and guanylate cyclase (IC50 is 4 μM). It inhibits cholera toxin- and Escherichia coli (STa) toxin- induced CFTR chloride current, ameliorates secretory diarrhea in mouse models, and prevents cyst growth in polycystic kidney disease model.
  • Inhibitor for the elevation of cAMP and cGMP induced by agonist.
  • Inhibits cholera toxin- and Escherichia coli (STa) toxin- induced CFTR chloride current.
  • Ameliorates secretory diarrhea in mouse models.
  • Prevents cyst growth in polycystic kidney disease model.