Learn More
Medchemexpress LLC Endo CNTinh-03 | 345288-49-5 | 427.51 | 5 MG
SDP
Catalog No. 5000401431
Encompass_Preferred
Endo CNTinh-03 is an inhibitor designed to prevent the elevation of cyclic AMP (cAMP) and cyclic GMP (cGMP) that are typically induced by agonists like G protein-coupled receptors, adenylate cyclase, and guanylate cyclase.
  • Inhibits cholera toxin-induced CFTR chloride current
  • Inhibits Escherichia coli (STa) toxin-induced CFTR chloride current
  • Ameliorates secretory diarrhea in mouse models
  • Prevents cyst growth in polycystic kidney disease models
  • Exhibits an IC50 of 4 μM against the elevation of cAMP and cGMP
Catalog No. 50-004-01431 Supplier Medchemexpress LLC Supplier No. HY4019135MG
Endo CNTinh-03 is an inhibitor designed to prevent the elevation of cyclic AMP (cAMP) and cyclic GMP (cGMP) that are typically induced by agonists like G protein-coupled receptors, adenylate cyclase, and guanylate cyclase.
  • Inhibits cholera toxin-induced CFTR chloride current
  • Inhibits Escherichia coli (STa) toxin-induced CFTR chloride current
  • Ameliorates secretory diarrhea in mouse models
  • Prevents cyst growth in polycystic kidney disease models
  • Exhibits an IC50 of 4 μM against the elevation of cAMP and cGMP