Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

endo-DNTT-PMI (DNTT-Precursor) 97.0+%, TCI America™
SDP

Catalog No. D515350MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D515350MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D515350MG Supplier TCI America Supplier No. D515350MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1269669-42-2
Molecular Formula C32H19NO2S2
Synonym endo-Dinaphthothienothiophene-phenylmaleimide Monoadduct, endo-Naphtho[2,3-b]naphtho[2′,3′:4,5]thieno[2,3-d]thiophene N-Phenylmaleimide Monoadduct

Specifications

Quantity 50 mg
Formula Weight 513.63
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material endo-DNTT-PMI (DNTT-Precursor)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.