Learn More
Omega Biotek Inc Endo-Free Plasmid DNA Maxi Kit, 25 preps
Catalog No. NC0768143
Encompass
Endo-Free Plasmid DNA Maxi Kit provides an efficient method for the large-scale isolation of low-endotoxin plasmid DNA. The system utilizes Lysate Clearance Filter Syringes instead of centrifugation to rapidly clear bacterial lysates post-alkaline lysis. Following lysate clearance, endotoxins are removed, and the protocol follows a simple “bind-wash-elute” procedure to deliver high-quality plasmid DNA.
Catalog No. NC0768143 Supplier Omega Biotek Inc Supplier No. D692603
Endo-Free Plasmid DNA Maxi Kit provides an efficient method for the large-scale isolation of low-endotoxin plasmid DNA. The system utilizes Lysate Clearance Filter Syringes instead of centrifugation to rapidly clear bacterial lysates post-alkaline lysis. Following lysate clearance, endotoxins are removed, and the protocol follows a simple “bind-wash-elute” procedure to deliver high-quality plasmid DNA.