Learn More
Neuromics, Inc ENDO-Growth Media, 500 ml is a complete culture media designed for human endothelial cells. The media contains 475 of ENDO-Basal media and 25 ml of ENDO-Growth supplements mixed together.
Catalog No. NC2060132
Encompass
ENDO-Growth Media contains two components freshly mixed for your convenience: 475 ml of ENDO-Basal media and 25 ml of ENDO-Growth supplement. ENDO-Basal Media: Media containing central vitamins and amino acids, and 0.5% FBS. ENDO-Growth Supplement: Contains the essential nutrients for endothelial cell proliferation; including 5% fetal bovine serum, recombinant growth factors and 1x penicillin and streptomycin.
Catalog No. NC2060132 Supplier Neuromics, Inc Supplier No. MED001
ENDO-Growth Media contains two components freshly mixed for your convenience: 475 ml of ENDO-Basal media and 25 ml of ENDO-Growth supplement. ENDO-Basal Media: Media containing central vitamins and amino acids, and 0.5% FBS. ENDO-Growth Supplement: Contains the essential nutrients for endothelial cell proliferation; including 5% fetal bovine serum, recombinant growth factors and 1x penicillin and streptomycin.