Learn More
Neuromics, Inc ENDO-Growth Media Kit, 500 ml is an all in one culture media kit designed for human endothelial cells. The kit contains 475 of ENDO-Basal media and 25 ml of ENDO-Growth supplements.
Catalog No. NC2071911
Encompass
All in one culture media kit designed for Human Endothelial CellsENDO-Growth Media Kit contains two components: 475 ml of ENDO-Basal media and 25 ml of ENDO-Growth supplement. ENDO-Basal Media: Media containing central vitamins and amino acids, and 0.5% FBS.ENDO-Growth Supplement: Contains the essential nutrients for endothelial cell proliferation; including 5% fetal bovine serum, recombinant growth factors and 1x penicillin and streptomycin
Catalog No. NC2071911 Supplier Neuromics, Inc Supplier No. EGK001
All in one culture media kit designed for Human Endothelial CellsENDO-Growth Media Kit contains two components: 475 ml of ENDO-Basal media and 25 ml of ENDO-Growth supplement. ENDO-Basal Media: Media containing central vitamins and amino acids, and 0.5% FBS.ENDO-Growth Supplement: Contains the essential nutrients for endothelial cell proliferation; including 5% fetal bovine serum, recombinant growth factors and 1x penicillin and streptomycin