Learn More
Angio Proteomie Endothelial Growth Medium (EGM), CAP-02
SDP
Catalog No. NC0731665
Encompass
Angio-Proteomie's Endothelial Growth Medium (EGM) is a complete, serum-free medium for culturing human endothelial cells from various tissues, ensuring optimal growth, proliferation, and function. It includes amino acids, vitamins, hormones, growth factors, and minerals. The 500 ml EGM requires supplements and has a 12-month shelf life.
Catalog No. NC0731665 Supplier Angio Proteomie Supplier No. CAP02
Angio-Proteomie's Endothelial Growth Medium (EGM) is a complete, serum-free medium for culturing human endothelial cells from various tissues, ensuring optimal growth, proliferation, and function. It includes amino acids, vitamins, hormones, growth factors, and minerals. The 500 ml EGM requires supplements and has a 12-month shelf life.