Learn More
QA Bio Inc Endo H - 20 uL
Supplier:  QA Bio Inc EEH0220
Encompass
Endo H cleaves Asparagine-linked hybrid or high mannose oligosaccharides, but not complex oligosaccharides. It cleaves between the two N-acetylglucosamine residues in the diacetylchitobiose core of the oligosaccharide, generating a truncated sugar molecule with one N-acetylglucosamine residue remaining on the asparagine. In contrast, PNGase F removes the oligosaccharide intact. Detergent and heat denaturation may increase the rate of cleavage for some glycoproteins. • Source: recombinant from Streptomyces plicatus in E.Coli • EC 3.2.1.96 • Contents: 60 µl aliquot of enzyme (300 mU) in 20 mM Tris-HCl, 25 mM NaCl, 1 mM EDTA (pH 7.5). / 200 µl 5x Reaction Buffer 5.5 (250 mM sodium phosphate, pH 5.5) - Denaturation Solution – 2% SDS, 1 M Beta-mercaptoethanol • Specific Activity >40 U/mg • Activity >5 U/ml • Molecular weight 29,000 daltons
Catalog No. 50-223-6652