Learn More
ACROBiosystems Endoglycosidase H (Endo H) is a recombinant glycosidase cloned from Streptomyces plicatus and overexpressed in E. coli. Endo H cleaves the chitobiose core of high-mannose oligosaccharides and a limited number of hybrid oligosaccharides from
Catalog No. 502253101
Encompass_Preferred
Endoglycosidase H is a recombinant glycosidase which cleaves within the chitobiose core of high mannose and some hybrid oligosaccharides from N-linked glycoproteins. It is a recombinant glycosidase cloned from Streptomyces plicatus and overexpressed in E. coli.
This item is not returnable. View return policy
Catalog No. 50-225-3101 Supplier ACROBiosystems Supplier No. ENHS511650000U
Endoglycosidase H is a recombinant glycosidase which cleaves within the chitobiose core of high mannose and some hybrid oligosaccharides from N-linked glycoproteins. It is a recombinant glycosidase cloned from Streptomyces plicatus and overexpressed in E. coli.