Learn More
New England Biolabs, Inc. Endo Hf – 100000 units
SDP
Catalog No. 50811828
Encompass
Endo Hf is a recombinant protein fusion of Endoglycosidase H and maltose binding protein. Endo Hf cleaves within the chitobiose core of high mannose and some hybrid oligosaccharides from N-linked glycoproteins equally as well as Endo H.
Catalog No. 50-811-828 Supplier New England Biolabs, Inc. Supplier No. P0703S
Endo Hf is a recombinant protein fusion of Endoglycosidase H and maltose binding protein. Endo Hf cleaves within the chitobiose core of high mannose and some hybrid oligosaccharides from N-linked glycoproteins equally as well as Endo H.