Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

endo-(±)-Norborneol, 92%, Thermo Scientific™

Catalog No. AC181460100
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC181460100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC181460100 Supplier Thermo Scientific Chemicals Supplier No. 181460100

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 497-36-9
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00167747,MFCD00063677
InChI Key ZQTYQMYDIHMKQB-UHFFFAOYNA-N
IUPAC Name bicyclo[2.2.1]heptan-2-ol
SMILES OC1CC2CCC1C2

Specifications

Melting Point 148.0°C to 154.0°C
Quantity 10 g
Formula Weight 112.17
Chemical Name or Material endo-(±)-Norborneol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.