Learn More
Agilent Technologies ENDO-O-GLYCOSIDASE
Catalog No. NC9012868
Encompass
Releases unsubstituted, Ser/Thr-linked GalGalNAc from proteins; used with additional enzymes to remove more complex O-linked structures. Also known asEndo-O-Glycosidase, Endo- N-acetylgalactosaminidase, O-Glycosidase.
Catalog No. NC9012868 Supplier Agilent Technologies Supplier No. GK80090
Releases unsubstituted, Ser/Thr-linked GalGalNAc from proteins; used with additional enzymes to remove more complex O-linked structures. Also known asEndo-O-Glycosidase, Endo- N-acetylgalactosaminidase, O-Glycosidase.