Learn More
Gene Tools LLC Endo-Porter | DMSO formulation | 1mL
SDP
Catalog No. NC1719519
Encompass
Endo-Porter is a novel peptide* explicitly designed to do what all existing peptide or lipid-based delivery reagents can not: deliver substances into the cytosol of cells by an endocytosis-mediated process that avoids damaging the plasma membrane of the cell. This mechanism prevents the loss of vital cell contents with its associated toxicity. Endo-Porter requires no interaction with the cargo to be delivered and allows the cargo that is endocytosed along with Endo-Porter to exit the endosome and enter the cytosol.
Catalog No. NC1719519 Supplier Gene Tools LLC Supplier No. OTEPDMSO1
Endo-Porter is a novel peptide* explicitly designed to do what all existing peptide or lipid-based delivery reagents can not: deliver substances into the cytosol of cells by an endocytosis-mediated process that avoids damaging the plasma membrane of the cell. This mechanism prevents the loss of vital cell contents with its associated toxicity. Endo-Porter requires no interaction with the cargo to be delivered and allows the cargo that is endocytosed along with Endo-Porter to exit the endosome and enter the cytosol.